Learn More
DWK Life Sciences Kimble™ Kontes™ Reflux Condenser with Internal, Coil-Type Cold Finger
SDP
Tightly wound coil provides enough surface area to condense high vapor pressure solvents like hexane
Supplier:  DWK Life Sciences 2830000000
Catalog No. K283000-0000
Only null left
Add to Cart
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Case
14/20
Condense high vapor pressure solvents like hexane
Reflux Condenser
200 mm
7.87 in.
Borosilicate Glass
USP Type I and ASTM E438, Type I, Class A
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.