Learn More
DWK Life Sciences Kimble Chase™ Plain Form API ASTM™ Hydrometers
SDP
Uses API scale (American Petroleum Institute) to measure specific gravity of a liquid relative to water
Supplier:  DWK Life Sciences 521307991
 View more versions of this product
Catalog No. 10-310-341
Only null left
Add to Cart
Description
Description
  • Serial number printed on the scale
  • Ballast is made from steel pellets and a binder, and is free of heavier metals
  • Includes Certificate of Traceability
  • Certificate of Traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
  • Meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plain Form API ASTM Hydrometer
0.10° API
Certificate of Traceability
25 mm
600 mL
9H
330 mm
7 mm
79 to 91° API
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.