Learn More
DWK Life Sciences Kimble Chase™ Plain Form API ASTM™ Hydrometers
SDP
Uses API scale (American Petroleum Institute) to measure specific gravity of a liquid relative to water
$62.25 - $72.70
Specifications
Type Plain Form API ASTM Hydrometer
Increments 0.10° API
Includes Certificate of Traceability
Volume (Metric) 600 mL
View More Specs
Products 10
Catalog Number Mfr. No. Specific Gravity Range ASTM Number Diameter (Metric) Outer Stem Diameter (Metric) Outside Price Quantity & Availability  
Catalog Number Mfr. No. Specific Gravity Range ASTM Number Diameter (Metric) Outer Stem Diameter (Metric) Outside Price Quantity & Availability  
10-310-335
SDP
View Documents
DWK Life Sciences
521301931
19 to 31° API 3H 8 mm 25 mm
Case for $72.70
Only null left
Add to Cart
10-310-336
SDP
View Documents
DWK Life Sciences
521302941
29 to 41° API 4H 7 mm 25 mm
Case for $62.50
Only null left
Add to Cart
10-310-337
SDP
View Documents
DWK Life Sciences
521303951
39 to 51° API 5H 7 mm 25 mm
Case for $64.25
Only null left
Add to Cart
10-310-338
SDP
View Documents
DWK Life Sciences
521304961
49 to 61° API 6H 7 mm 25 mm
Case for $66.10
Only null left
Add to Cart
10-310-339
SDP
View Documents
DWK Life Sciences
521305971
59 to 71° API 7H 7 mm 25 mm
Case for $66.60
Only null left
Add to Cart
10-310-340
SDP
View Documents
DWK Life Sciences
521306981
69 to 81° API 8H 7 mm 25 mm
Case for $66.60
Only null left
Add to Cart
10-310-341
SDP
View Documents
DWK Life Sciences
521307991
79 to 91° API 9H 7 mm 25 mm
Case for $62.25
Only null left
Add to Cart
10-310-342
SDP
View Documents
DWK Life Sciences
521308910
89 to 101° API 10H 5 mm 21 mm
Case for $64.25
Only null left
Add to Cart
10-310-343
SDP
View Documents
DWK Life Sciences
521300111
-1 to 11° API 1H 8 mm 25 mm
Case for $65.75
Only null left
Add to Cart
10-310-344
SDP
View Documents
DWK Life Sciences
521300921
9 to 21° API 2H 8 mm 25 mm
Case for $63.75
Only null left
Add to Cart
Description
Description
  • Serial number printed on the scale
  • Ballast is made from steel pellets and a binder, and is free of heavier metals
  • Includes Certificate of Traceability
  • Certificate of Traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
  • Meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
Specifications
Specifications
Plain Form API ASTM Hydrometer
0.10° API
Certificate of Traceability
600 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.