Learn More
DWK Life Sciences Kimble Chase™ Precision Specific Gravity Hydrometers
SDP
Available in a variety of graduation ranges and overall lengths
Supplier:  DWK Life Sciences 521100910
 View more versions of this product
Catalog No. 13-757-037
Only null left
Add to Cart
Description
Description
  • Serial number printed on the scale
  • Ballast is made from steel pellets and a binder, and is free of heavier metals
  • Includes Certificate of Traceability
  • Certificate of Traceability indicates that the hydrometers are manufactured using devices calibrated using NIST traceable standards
  • Meets or exceeds the requirements in ASTM specification E100, and it is calibrated to ASTM specification E126 to ensure accuracy
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Precision Specific Gravity Hydrometer
310 mm
7 mm
0.900 to 1.010
0.001 sp.gr.
Certificate of Traceability
18 mm
500 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.