Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
SDP
Catalog No. 0334014B Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03-340-14B 18-400
03-340-14D 8-425
03-340-14F 13-425
03-340-14E 15-425
03-340-14C 20-400
03-340-14A 24-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 03-340-14B Supplier DWK Life Sciences Supplier No. 7380218400
Add to Cart
Add to Cart
This item is not returnable. View return policy
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
TRUSTED_SUSTAINABILITY
Specifications
Closure Size 18-400
Liner Material PTFE, Rubber
Product Type Screw Cap
Material Phenolic
Color Black