Learn More
DWK Life Sciences Kimble™ Closures for Sample Vials
GSA_VASDP
Catalog No. 0334014C Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03-340-14C 20-400
03-340-14D 8-425
03-340-14B 18-400
03-340-14F 13-425
03-340-14E 15-425
03-340-14A 24-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 03-340-14C Supplier DWK Life Sciences Supplier No. 7380220400
Add to Cart
Add to Cart
Black phenolic closure with PTFE-lined cap
  • Works with media bottles and screw-thread vials
  • Special black phenolic material (wood filled) very resistant to the effects of temperature and steam encountered in autoclaving
  • Cemented-in PTFE-faced rubber liner is provided
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Screw Cap
Closure Size 20-400
Liner Material PTFE, Rubber
Material Phenolic
Color Black