Learn More
DWK Life Sciences Kimble™ Enlarging Connecting Adapters
Shop All DWK Life Sciences Products
Click to view available options
Standard Taper Joint No.:
Outer Top: 10/30, Inner Bottom: 24/40
Description
- Outer joint at top, inner at bottom
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type 1 and ASTM E438, Type 1, Class A requirements
Specifications
Specifications
For Use With (Application) | For enlarging size of Standard Taper glass joints |
Material | Borosilicate Glass (33 Low Extractable) |
Product Type | Enlarging Connecting Adapter |
Standard Taper Joint No. | Outer Top: 10/30, Inner Bottom: 24/40 |