Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Reagent Sprayers with Screw Thread Ground Joint
GSA_VASDP

Catalog No. p-73088 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
10 mL
50 mL
125 mL
250 mL
Diameter (Metric) Outer:
2.2 cm
5 cm
6.5 cm
8.5 cm
Standard Taper Joint:
19/22
24/40

Provide uniform spray mist of visualization reagents for TLC and paper chromatography; use with all standard TLC reagents and most corrosive liquids

  • Liquid touches only nonreactive borosilicate glass atomizing sprayer head and flask
  • Sprayer head-to-flask connection consists of screw-thread standard taper ground joint, connecting cap, and loosening ring
  • Operate sprayer at 3 to 5psi (0.021 to 0.035MPa)
  • Adjust spray pattern by varying the position of thumb on vent hole
  • Autoclavable (after cap, loosening ring, and O-ring are removed)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Capacity (English) 8.45 oz.
Capacity (Metric) 250 mL
Connections Screw thread ground joint, connecting cap, loosening ring
Material Glass
Quantity 1 Case
Type TLC Reagent Sprayer
For Use With (Application) To Provide Uniform Spray Mist of Visualization Reagents for TLC and Paper Chromatography
For Use With (Equipment) Standard TLC Reagents and Most Corrosive Liquids
Operating Pressure 3 to 5 psi
Standard Taper Joint 24/40
Height (English) 9.01 in.
Height (Metric) 22.9 cm
Diameter (English) Outer 3.34 in.
Diameter (Metric) Outer 8.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.