Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Solvent Filtration Assemblies
SDP

Catalog No. K9538201047 Shop All DWK Life Sciences Products
Click to view available options
Volume (Metric):
1 L
4 L
Height (Metric):
25.4 cm
52.5 cm

Includes

  • Solvent pickup adapter
  • Coarse porosity fritted glass filter support with No. 2 vacuum hose connection
  • Anodized aluminum clamp
  • Filter flask

Eliminate pouring HPLC solvents into funnels

  • Draw solvent directly from reagent bottle through 47mm filter and into filter flask
  • After filtration, stopcock on adapter can be turned off for vacuum degassing
  • Solvent pickup adapter is available separately to convert funnel assemblies

TRUSTED_SUSTAINABILITY

Specifications

Material Glass
Type Flask
Volume (Metric) 1 L
Width (English) 8.0625 in.
Width (Metric) 20.5 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.