Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Kontes™ Ultra-Ware™ Solvent Filtration Assemblies
Shop All DWK Life Sciences Products

Click to view available options
Volume (Metric):
1 L
4 L
Height (Metric):
25.4 cm
52.5 cm
Description
Includes
- Solvent pickup adapter
- Coarse porosity fritted glass filter support with No. 2 vacuum hose connection
- Anodized aluminum clamp
- Filter flask
- Draw solvent directly from reagent bottle through 47mm filter and into filter flask
- After filtration, stopcock on adapter can be turned off for vacuum degassing
- Solvent pickup adapter is available separately to convert funnel assemblies
Specifications
Specifications
| Material | Glass |
| Type | Flask |
| Volume (Metric) | 1 L |
| Width (English) | 8.0625 in. |
| Width (Metric) | 20.5 cm |
| Height (English) | 10 in. |
| Height (Metric) | 25.4 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction