Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Open-Top Aluminum Seals, Color Coded
SDP

Catalog No. 03340305 Shop All DWK Life Sciences Products
Click to view available options
Color:
Blue
Green
Natural
Red
Closure Size:
13 mm
20 mm

Designed to secure septa and stoppers to bottles and vials

  • For use with 13 and 20mm serum finished bottles and vials
  • Available in blue, red, green and natural aluminum color to help separate and identify samples

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
Material Aluminum
Quantity 1000 Cs.
Type Open Top Cap
Closure Size 13 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.