Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Solid PE Liners
SDP

Catalog No. 13757133
Click to view available options
Neck Finish:
20-400
22-400
24-400
28-400
33-400
38-400
43-400
45-400
48-400
53-400
58-400
63-400
70-400
89-400
Not recommended for hydrocarbon solvents.
Not recommended for hydrocarbon solvents.

Has good resistance to acids, alcohols, alkalis, aqueous solutions, oils, solvents

  • General purpose phenolic screw cap with a solid polyethylene (PE) liner
  • Color: black
  • Liner material: solid polyethylene (0.040-in.)
  • Ideal for use with strong acid or base samples
  • Not recommended for hydrocarbon solvents

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 43-400
Product Type Phenolic Cap
For Use With (Equipment) Bottle
Closure Liner Polyethylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.