Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences KIMBLE™ Silicone Lids
SDP

Catalog No. 15112181 Shop All DWK Life Sciences Products
Change view
Click to view available options
Color:
Cyan
Green
Pink
Description:
Large
Medium
Small
Small, Medium & Large
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Description
15-112-181 Cyan Small, Medium & Large
15-112-171 Pink Small
15-112-172 Cyan Small
15-112-173 Green Small
15-112-174 Pink Medium
15-112-175 Cyan Medium
15-112-176 Green Medium
15-112-177 Pink Large
15-112-178 Cyan Large
15-112-179 Green Large
15-112-180 Pink Small, Medium & Large
15-112-182 Green Small, Medium & Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 15-112-181 Supplier DWK Life Sciences Supplier No. 291110029
Only null left
Add to Cart
Add to Cart

Reusable lids stretch easily and adapt to many container sizes, improves lab safety, enhances sample management, offers a more sustainable and cost-effective solution over foils, films and other single-use covers.

VERSATILE

  • Fits on beakers, flasks, bottles, cylinders, plastic and glass products
  • Each size conforms to various diameters
  • Works with spouts, beads and thread finishes
  • Adaptable for many uses beyond limits of traditional films and covers

SAMPLE MANAGEMENT

  • Reduces cross-contamination of samples
  • Compatible with lab markers to enhance sample identification
  • Improves your lab workflow efficiency
  • Uses color to track projects and samples

LAB SAFETY

  • Improves safety performance over foils or films
  • Color aids quick visual identification
  • Protects skin and nose from contents
  • Defends against spill/leakage

SUSTAINABLE

  • Reusable, customizable and dishwasher safe
  • Reduces non-reusable consumables that generate waste such as films and foils
  • Chemically resistant, autoclavable and pliable over a wide temperature range
  • Reduces environmental impact and lowers lab operational costs
TRUSTED_SUSTAINABILITY

Specifications

Color Cyan
Description Small, Medium & Large
Diameter (Metric) Outer 43 to 61 mm (small), 64 to 76 mm (medium), 84 to 116 mm (large)
Material Silicone
For Use With (Application) To cover open lab vessels
For Use With (Equipment) Beakers, Bottles, Cylinders, Erlenmeyer Flask
Height (Metric) 22.5 mm (small), 22.5 mm (medium), 23.2 mm (large)
Length (Metric) 16.00 cm
Quantity 3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.