Learn More
DWK Life Sciences™ Kimble™ Stainless Steel Standard Taper Joint Clamp
SDP
Catalog No. p-7240073 Shop All DWK Life Sciences Products
Click to view available options
Standard Taper Joint:
14/20, 14/35
This item is not returnable. View return policy
This item is not returnable. View return policy
The Kimble™ Stainless Steel Standard Taper Joint Clamp is used to secure standard ground joint connections in glass assemblies and is characterized by its easy handling and excellent resilience.
  • Available in either metal or polyoxymethylene (POM)
  • Dimensionally stable with particularly good spring properties
  • Easy, safe and quick to fit
  • Superior chemical and temperature resistance
  • Can be heated up to 500°C
  • Plastic clips are marked with their joint size and are color coded to allow easy identification of the size of clip required
TRUSTED_SUSTAINABILITY
Specifications
Type Standard Taper Joint Clamp
Material Stainless Steel
Standard Taper Joint 14/20, 14/35
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.