Learn More
Ansell Kimtech™ A8 Zippered Lab Coats
GREENER_CHOICE
Catalog No. 19005115 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
XL
2XL
4XL
5XL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-115 XL
19-005-112 Small
19-005-113 Medium
19-005-114 Large
19-005-116 2XL
19-005-117 4XL
19-005-118 5XL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-005-115 Supplier Ansell Supplier No. 51934
Add to Cart
Add to Cart
This item is not returnable. View return policy
Kimtech™ A8 Zippered Lab Coats are made with a protective 3-layer SMS fabric and are recommended where there is the potential for light fluid contact.
  • Zippered front closure and no pockets to reduce risk of contamination
  • Soft knit cuffs
  • High mandarin collar
  • Double bagged
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Lab Coat
Size XL
Gender Unisex
Material SMS
Color White
Closure Type Zipper
Cuff Style Knit
Disposable Limited-use
Sterility Non-sterile
No. of Pockets None
Collar Type Mandarin
Model 51934
No. per Pack 25/Cs.
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.