Learn More
Ansell Kimtech™ N95 Pouch Respirator
GREENER_CHOICE
N95 pouch respirator is NIOSH-Approved and provides a minimum 95% filtration efficiency of 0.3 micron particles.
$582.67
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color White
Design Type Pouch/Duckbill
Disposable Single-use
View More Specs
Alert:
Not for medical use. Not to be used by healthcare professionals or in healthcare settings.
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-005-123
GREENER_CHOICE
View Documents
Ansell
53358
One Size
Case of 300 for $582.67
Only null left
Edge
Add to Cart
17-986-014
View Documents
Ansell
54066
Small N/A N/A N/A
Description
Description
  • NIOSH-Approved, provides a minimum 95% filtration efficiency of 0.3 micron particles
  • Guards against the risks posed by airborne particles and contaminants
  • Pouch-style offers the wearer a large breathing chamber to maximize comfort
  • Equipped with two comfort fit headbands and a bendable nosepiece to ensure a consistent seal
  • Not intended for the healthcare market and/or healthcare personnel
  • 300 Kimtech™ N95 Pouch Respirators (6 bags of 50)
Specifications
Specifications
N - Not Resistant to Oil
White
Single-use
General Purpose Particulate Respirator
95%
Non-sterile
No
NIOSH 42 CFR 84, N95
Pouch/Duckbill
N95
No
Adjustable Nose Clip
Two Comfort Fit Headbands
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.