Description
- Presented unzipped with the inside of the garment first
- Arms telescope folded
- Legs telescope folded with built-in snaps
- Elastic cuffs and thumb loops
- No label on the outer surface
- Hood, front panel, arms and leg made of high performance laminated SMS fabric
- Back panel made of ultralight weight breathable SMS
- 1 large bag of 25 coveralls individually packed per case
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | X-Large |
| Material | SMS |
| Disposable | Single-use |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Sterility | Non-sterile |
| Certifications/Compliance | PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection; EN 14126:2003 certified protection against infective agent penetration |
| For Use With (Application) | Controlled Environments |
| Cleanroom Class | Class 10,000-100,000 (ISO 7-8) |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.