Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ A8 Cleanroom Coveralls
GREENER_CHOICE

Catalog No. 19169222F Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-222F 3X-Large
19-169-222A Small
19-169-222B Medium
19-169-222C Large
19-169-222D X-Large
19-169-222E 2X-Large
19-169-222G 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-169-222F Supplier Ansell Supplier No. 47956
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cleanroom coveralls are designed to enhance comfort and reduce heat stress in ISO 7 and 8 environments.

Kimtech™ A8 Cleanroom Coveralls are made with a lightweight, innovative fabric that moves with the worker, and feature built-in thumb loops to protect your people and processes, and a high neck collar to control contamination risk. These white coveralls also have elastic wrists, ankles and waist.

They are designed for protection against dust, chemicals and other contaminants and have a BFE of 94%. The white coveralls are designed for applications including ISO 7/8 cleanrooms, Grade C/D, non-sterile pharmaceutical manufacturing, bio-medical research, pharmaceutical compounding and biotechnology.

Plus, these protective suits are recyclable through the RightCycle Program.

  • 25 Kimtech™ A8 Coveralls/Case
  • Helps protect against dust, chemicals and other contaminants; BFE of 94%; ISO 7/8 cleanrooms
  • Low lint for contamination control
  • Recyclable through the RightCycle Program, which turns used lab coats into raw materials for eco-friendly consumer products
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Coveralls
Size 3X-Large
Material SMS
Color White
Disposable Single-use
Cuff Style Thumb Loop
Neck Style Collared
Sterility Non-sterile
Resistance Anti-static
Collar Type Standing
Certifications/Compliance PPE Cat III directive 89/686/EEC, Type 6 limited chemical splash protection; Type 5 particle protection
Model Kimtech™ A8
For Use With (Application) Pharmaceutical, Packing, Distribution, Production, Biotechnology
Packaging Double Bagged
Cleanroom Class Class 10,000-100,000 (ISO 7-8)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.