Learn More
Fisherbrand™ Disposable Face Shields
SDPSCI_ED
Catalog No. S98127 Shop All Fisherbrand Products
Change view
Click to view available options
Model:
Full face
Catalog No. Model
S98127 Full face
1 options
This item is not returnable. View return policy
Catalog No. S98127
Add to Cart
Add to Cart
This item is not returnable. View return policy
High-quality, optically clear, distortion-free 7mil film.
  • Polyester lens is treated with antifog and antistatic coating
  • Headpiece is held in place by a stretch strap and includes a latex-free foam band for comfortable extended wear
Compliance
Conform to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendations for protective eyewear.
Specifications
Thickness (English) 7 mil
Lens Coating Anti-fog, Anti-static
Lens Tint Clear
Includes Stretch Strap, Latex-free Foam Band
Model Full face
Size X-Large
Material Polyester
Disposable Single-use
Lens Material Polyester