Learn More
Fisherbrand™ Disposable Face Shields
SDPSCI_ED
High-quality, optically clear, distortion-free 7mil film.
$71.20
Specifications
Lens Coating Anti-fog, Anti-static
Thickness (English) 7 mil
Lens Tint Clear
Includes Stretch Strap, Latex-free Foam Band
Material Polyester
View More Specs
Products 1
Catalog Number Model Price Quantity & Availability  
Catalog Number Model Price Quantity & Availability  
S98127
SDPSCI_ED
View Documents
Full face
Pack of 25 for $71.20
Case of 4 PK for $285.00
Only null left
Edge
Add to Cart
Description
Description
  • Polyester lens is treated with antifog and antistatic coating
  • Headpiece is held in place by a stretch strap and includes a latex-free foam band for comfortable extended wear
Compliance
Conform to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendations for protective eyewear.
Specifications
Specifications
Anti-fog, Anti-static
Clear
Polyester
Polyester
7 mil
Stretch Strap, Latex-free Foam Band
Single-use
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.