Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579556
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579556 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579556 Supplier Invitrogen™ Supplier No. PA579556
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Skeletal Muscle Tissue, Human Placenta Tissue, Rat Testis Tissue.

Kin is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by the human ortholog.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIN
Applications Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal
Gene Kin
Gene Accession No. O60870, Q8K339
Gene Alias antigenic determinant of rec-A protein; antigenic determinant of rec-A protein homolog; binding to curved DNA; Btcd; DNA/RNA-binding protein KIN17; HsKin17 protein; KIN; KIN, antigenic determinant of recA protein; KIN, antigenic determinant of recA protein homolog; KIN17; Kin17 DNA and RNA binding protein; Rts2
Gene Symbols Kin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence at the N-terminus of human KIN (48-64aa ESHQRQLLLASENPQQF).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16588, 22944, 689197
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.