Learn More
Invitrogen™ KIN Polyclonal Antibody

Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell, human 293T whole cell, human HepG2 whole cell, rat RH35 whole cell, rat NRK whole cell, mouse HEPA1-6 whole cell, mouse NIH/3T3 whole cell. IHC: human oesophagus squama cancer tissue.
Specifications
Specifications
| Antigen | KIN |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5mg BSA and 0.01mg sodium azide |
| Gene | Kin |
| Gene Accession No. | O60870, Q8K339 |
| Gene Alias | antigenic determinant of rec-A protein; antigenic determinant of rec-A protein homolog; binding to curved DNA; Btcd; DNA/RNA-binding protein KIN17; HsKin17 protein; KIN; KIN, antigenic determinant of recA protein; KIN, antigenic determinant of recA protein homolog; KIN17; Kin17 DNA and RNA binding protein; Rts2 |
| Gene Symbols | Kin |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.