Learn More
Thermo Scientific™ Kinase buffer I (5X)
For cellular and molecular biology applications
$62.90 - $83.10
Specifications
Chemical Name or Material | Kinase buffer I (5X) |
---|---|
Physical Form | Liquid |
Formulation | 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ61967AK
|
Thermo Scientific™
J61967AK |
250 mL |
Each for $83.10
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ61967AE
|
Thermo Scientific™
J61967AE |
100 mL |
Each for $62.90
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Formulation: 100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4
Specifications
Kinase buffer I (5X) | |
Liquid | |
100mM Tris-HCl 100, 50mM magnesium chloride, 5mM dithiothreitol, and 5mM sodium orthovanadate, pH 7.4. |
Safety and Handling
Recommended Storage : Ambient temperatures
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.