Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Kinase Selectivity Profiling System: TK-3 + ADP-Glo™ Assay

Catalog No. PRV6921 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
8 x 50 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV6921 8 x 50 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV6921 Supplier Promega Supplier No. V6921
Only null left
Add to Cart
Add to Cart

Easy-to-use kits for performing kinase selectivity profiling that rely on the ADP-Glo™ Kinase Assay technology. Each system includes kinase and substrate pairs in an easy-to-use 8-tube strip format optimized for fast and simple kinase profiling reactions.

  • Selectivity Screening of Library Compounds for Effect on Tyrosine Kinases
  • Kinase and substrate pairs organized in an easy-to-use 8-tube strip format
  • Simple reaction assembly and automation-friendly protocol
  • Streamilines profiling of hits and leads for potency against families of related kinases

Specifications

Quantity 8 x 50 Reactions
Content And Storage Components at different temperatures
Format 384-well Plate
For Use With (Application)
  • Selectivity screening of library compounds for effects on target kinases
  • Profile hits and leads for their potency against families of related kinases
Kit Contents Cat.# V6920 Kinase Selectivity Profiling System: TK-3 which includes:
  • 2 TK-3 Kinase Strip
  • 2 TK-3 Substrate/Co-Factor Strip
  • 1.5mL 5X Reaction Buffer A
  • 25μL DTT, 0.1M
Cat.# V6930 ADP-Glo™ Kinase Assay, which includes:
  • 2mL ADP-Glo™ Reagent
  • 4mL Kinase Detection Reagent
  • 500μL Ultra Pure ATP, 10mM
  • 500μL ADP, 10mM
Product Type Kinase Selectivity Profiling System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.