Learn More
Invitrogen™ Kindlin 2 Monoclonal Antibody (OTI25A4)

Description
Kindlin 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Specifications
Specifications
| Antigen | Kindlin 2 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI25A4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | FERMT2 |
| Gene Accession No. | Q96AC1 |
| Gene Alias | AA960555; EGK_18193; Fermitin family homolog 2; fermitin family homolog 2 (Drosophila); fermitin family member 2; fermt2; im:7145859; im:7145859 protein; KIND2; kindlin 2; kindlin-2; LOC616981 protein; MIG2; mig-2; mitogen inducible 2; mitogen inducible gene 2 protein; mitogen-inducible gene 2 protein; PH domain-containing family C member 1; pleckstrin homology domain containing, family C (with FERM domain) member 1; pleckstrin homology domain containing, family C member 1; pleckstrin homology domain-containing family C member 1; Plekhc1; UNC112; UNC112B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.