Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kindlin 2 Monoclonal Antibody (OTI9E4)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524937
Description
Kindlin 2 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Scaffolding protein that enhances integrin activation mediated by TLN1 and/or TLN2, but activates integrins only weakly by itself. Binds to membranes enriched in phosphoinositides. Enhances integrin-mediated cell adhesion onto the extracellular matrix and cell spreading; this requires both its ability to interact with integrins and with phospholipid membranes. Required for the assembly of focal adhesions. Participates in the connection between extracellular matrix adhesion sites and the actin cytoskeleton and also in the orchestration of actin assembly and cell shape modulation. Recruits FBLIM1 to focal adhesions. Plays a role in the TGFB1 and integrin signaling pathways. Stabilizes active CTNNB1 and plays a role in the regulation of transcription mediated by CTNNB1 and TCF7L2/TCF4 and in Wnt signaling. [UniProt].
Specifications
| Kindlin 2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CIB5, Q96AC1 | |
| FERMT2 | |
| Full-length protein expressed in 293T cell transfected with human FERMT2 expression vector. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| OTI9E4 | |
| Unconjugated | |
| FERMT2 | |
| AA960555; EGK_18193; Fermitin family homolog 2; fermitin family homolog 2 (Drosophila); fermitin family member 2; fermt2; im:7145859; im:7145859 protein; KIND2; kindlin 2; kindlin-2; LOC616981 protein; MIG2; mig-2; mitogen inducible 2; mitogen inducible gene 2 protein; mitogen-inducible gene 2 protein; PH domain-containing family C member 1; pleckstrin homology domain containing, family C (with FERM domain) member 1; pleckstrin homology domain containing, family C member 1; pleckstrin homology domain-containing family C member 1; Plekhc1; UNC112; UNC112B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10979, 218952, 289992, 490693 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction