Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kindlin 2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559200 Supplier Invitrogen™ Supplier No. PA559200
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADSSYNLEV QNILSFLKMQ HLNPDPQLIP EQITTDITPE CLVSPRYLKK YKNKQITARI LEA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 97%.

Scaffolding protein that enhances integrin activation mediated by TLN1 and/or TLN2, but activates integrins only weakly by itself. Binds to membranes enriched in phosphoinositides. Enhances integrin-mediated cell adhesion onto the extracellular matrix and cell spreading; this requires both its ability to interact with integrins and with phospholipid membranes. Required for the assembly of focal adhesions. Participates in the connection between extracellular matrix adhesion sites and the actin cytoskeleton and also in the orchestration of actin assembly and cell shape modulation. Recruits FBLIM1 to focal adhesions. Plays a role in the TGFB1 and integrin signaling pathways. Stabilizes active CTNNB1 and plays a role in the regulation of transcription mediated by CTNNB1 and TCF7L2/TCF4 and in Wnt signaling. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kindlin 2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FERMT2
Gene Accession No. Q96AC1
Gene Alias AA960555; EGK_18193; Fermitin family homolog 2; fermitin family homolog 2 (Drosophila); fermitin family member 2; fermt2; im:7145859; im:7145859 protein; KIND2; kindlin 2; kindlin-2; LOC616981 protein; MIG2; mig-2; mitogen inducible 2; mitogen inducible gene 2 protein; mitogen-inducible gene 2 protein; PH domain-containing family C member 1; pleckstrin homology domain containing, family C (with FERM domain) member 1; pleckstrin homology domain containing, family C member 1; pleckstrin homology domain-containing family C member 1; Plekhc1; UNC112; UNC112B
Gene Symbols FERMT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Kindlin 2. Recombinant protein control fragment (Product #RP-98347).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.