Learn More
Invitrogen™ Kindlin 3 Monoclonal Antibody (OTI2G2)

Description
Kindlin 3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | Kindlin 3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2G2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Fermt3 |
| Gene Accession No. | Q86UX7 |
| Gene Alias | C79673; fermitin family homolog 3; fermitin family homolog 3 (Drosophila); fermitin family member 3; Fermt3; Kind3; kindlin 3; Kindlin3; Kindlin-3; MGC10966; MIG-2; MIG2B; MIG2-like protein; RGD1310168; UNC-112 related protein 2; UNC112C; unc-112-related protein 2; URP2; URP2SF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.