Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kindlin 3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566753 Supplier Invitrogen™ Supplier No. PA566753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MADSSYTSEVQ AILAFLSLQR TGSGGPGNHP HGPDASAEGL NPYGLVAPRF QRKFKAKQLT Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

The three KINDLINs are a novel family of focal adhesion proteins, localizing to integrin adhesion sites. The KINDLIN proteins are composed of a centrally located FERM domain interrupted by a pleckstrin homology (PH) domain. KINDLIN1 and KINDLIN2 have been shown to play an essential role in integrin-mediated adhesion and spreading. In contrast to the widely expressed KINDLIN1 and KINDLIN2, KINDLIN3 is restricted to hematopoietic cells and is particularly abundant in megakaryocytes and platelets. Several reports describe a transcriptional misregulation of KINDLINs in various types of cancer. A recent study demonstrates that KINDLIN3 is essential for platelet integrin activation and subsequent integrin outside-in signaling, suggesting it may serve as a potential target for the design of therapeutics aimed at specifically disrupting integrin activation in platelets and leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kindlin 3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fermt3
Gene Accession No. Q86UX7
Gene Alias C79673; fermitin family homolog 3; fermitin family homolog 3 (Drosophila); fermitin family member 3; Fermt3; Kind3; kindlin 3; Kindlin3; Kindlin-3; MGC10966; MIG-2; MIG2B; MIG2-like protein; RGD1310168; UNC-112 related protein 2; UNC112C; unc-112-related protein 2; URP2; URP2SF
Gene Symbols Fermt3
Host Species Rabbit
Immunogen Recombinant Human Kindlin 3. Recombinant protein control fragment (Product #RP-107411).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83706
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.