Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kindlin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519817
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519817 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519817 Supplier Invitrogen™ Supplier No. PA519817
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with cow and Xenopus laevis based on sequence homology.

Kindlin 1 is a member of the kindlin family of focal adhesion proteins and contains a FERM domain and pleckstrin homology domain. It is involved in integrin signaling, linkage of the cytoskeleton to the extracellular matrix, and cell adhesion. It is most often expressed in epithelial cells. When kindlin 1 is coexpressed with talin it helps to activate ITGA2B. It is required for keratinocyte proliferation, polarization of basal keratinocytes in the skin, and for normal cell shape. Mutations in kindlin 1 have been linked to Kindler syndrome, a recessive skin disorder involving blistering, photosensitivity, poikiloderma, and atrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kindlin
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene FERMT1
Gene Accession No. P59113, Q9BQL6
Gene Alias 5830467P10Rik; C20orf42; DTGCU2; Fermitin family homolog 1; fermitin family homolog 1 (Drosophila); fermitin family member 1; FERMT1; FLJ20116; FLJ23423; KIND1; Kindlerin; kindlin 1; kindlin syndrome protein; Kindlin-1; RGD1306816; RP5-1056H1.1; UNC112 related protein 1; UNC-112 related protein 1; UNC112A; unc-112-related protein 1; URP1
Gene Symbols FERMT1
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 650 to the C-terminus of Human Kindlin.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 241639, 55612
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.