Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kindlin Recombinant Rabbit Monoclonal Antibody (7H7L9)

Catalog No. 701830
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-183-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-183-0 Supplier Thermo Scientific Supplier No. 701830
Add to Cart
Edge
Add to Cart

Kindlin Recombinant Rabbit Monoclonal Antibody (7H7L9)

Kindlin Recombinant Monoclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen Kindlin
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7H7L9
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene FERMT1
Gene Accession No. Q9BQL6
Gene Alias 5830467P10Rik; C20orf42; DTGCU2; Fermitin family homolog 1; fermitin family homolog 1 (Drosophila); fermitin family member 1; FERMT1; FLJ20116; FLJ23423; KIND1; Kindlerin; kindlin 1; kindlin syndrome protein; Kindlin-1; RGD1306816; RP5-1056H1.1; UNC112 related protein 1; UNC-112 related protein 1; UNC112A; unc-112-related protein 1; URP1
Gene Symbols FERMT1
Host Species Rabbit
Immunogen Protein corresponding to Human FERMT1 (aa 200-409).
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55612
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.