Learn More
Kinectin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304384AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin (see MIM 148760 and MIM 600025), an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.Specifications
Kinectin | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q86UP2 | |
KTN1 | |
Between 1307 and 1357 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KTN1 | |
CG1, KNT, MU-RMS-40.19, CG-1 antigen, kinectin, kinesin receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3895 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.