Learn More
Kinematica HMC Europe GLP Set for HV-Series Autoclaves
Catalog No. 08451436 Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
HMC Autoclaves, HV-Series
Combines two frequently ordered accessories: Printer and Exhaust Air Filter.
Set includes:
  • Printer for recording sterilization (data, temperature and time) 
  • Exhaust air filter (inclusive filter cartridge) for sterilizing high infectious materials
  • In-line-sterilization cartridge
Note:  Image shows the HV Autoclave family.
Specifications
For Use With (Equipment) HMC Autoclaves, HV-Series
Product Type GLP Package
Includes Exhaust Air Filter, Cartridge and Printer
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.