Learn More
Kinematica HMC Europe Vertical Autoclave Temperature Sensors
Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
HMC-HG autoclave
Description
- Temperature sensing probe located inside the chamber.
- Designed so that the user can place the tip of the probe into the load being sterilized. It is usually placed in the most difficult to sterilize location.
Specifications
Specifications
For Use With (Equipment) | HMC-HG autoclave |
Product Type | Flexible Temperature Sensor |