Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin 5A Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB265594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB265594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB265594 Supplier Novus Biologicals Supplier No. NBP232231AF488
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kinesin 5A Polyclonal antibody specifically detects Kinesin 5A in Human samples. It is validated for Western Blot

Specifications

Antigen Kinesin 5A
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias D12S1889, KIF5A variant protein, kinesin family member 5A, kinesin heavy chain isoform 5A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 982 to 1032 of human Kinesin Family Member 5A using the numbering given in entry NP_004975.2 (GeneID 3798).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers
Primary or Secondary Primary
Gene ID (Entrez) 3798
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.