Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5A Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232231AF700
Description
Kinesin 5A Polyclonal antibody specifically detects Kinesin 5A in Human samples. It is validated for Western BlotSpecifications
Kinesin 5A | |
Polyclonal | |
Western Blot | |
D12S1889, KIF5A variant protein, kinesin family member 5A, kinesin heavy chain isoform 5A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10 | |
The immunogen this antibody was made to, maps to a region between residue 982 to 1032 of human Kinesin Family Member 5A using the numbering given in entry NP_004975.2 (GeneID 3798). | |
0.1 mL | |
Cell Biology, Cellular Markers | |
3798 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction