Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kinesin 5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528143
Description
Recommended positive controls: IMR32, mouse brain. Predicted reactivity: Mouse (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.
Specifications
| Kinesin 5A | |
| Polyclonal | |
| Unconjugated | |
| KIF5A | |
| D10Bwg0738e; D12S1889; Khc; Kiaa4086; Kif5; Kif5a; kif5a {ECO:0000312; KIF5A variant protein; kinesin family member 5A; kinesin heavy chain isoform 5A; kinesin heavy chain isoform 5A; kinesin heavy chain; kinesin heavy chain neuron-specific 1; kinesin, heavy chain, neuron-specific; Kns; mKIAA4086; MY050; Neuronal kinesin heavy chain; NKHC; Nkhc1; RGD:1303035}; SPG10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16572, 314906, 3798 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P33175, Q12840, Q6QLM7 | |
| KIF5A | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human KIF5A. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction