Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kinesin 5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529820
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529820 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529820 Supplier Invitrogen™ Supplier No. PA529820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, mouse brain, rat brain. Predicted reactivity: Mouse (95%), Rat (96%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin 5A
Applications Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KIF5A
Gene Accession No. P33175, Q12840, Q6QLM7
Gene Alias D10Bwg0738e; D12S1889; Khc; Kiaa4086; Kif5; Kif5a; kif5a {ECO:0000312; KIF5A variant protein; kinesin family member 5A; kinesin heavy chain isoform 5A; kinesin heavy chain isoform 5A; kinesin heavy chain; kinesin heavy chain neuron-specific 1; kinesin, heavy chain, neuron-specific; Kns; mKIAA4086; MY050; Neuronal kinesin heavy chain; NKHC; Nkhc1; RGD:1303035}; SPG10
Gene Symbols KIF5A
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human KIF5A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16572, 314906, 3798
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.