Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB500580
Description
Kinesin 5B Polyclonal specifically detects Kinesin 5B in Human, Mouse, Porcine samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Kinesin | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.4) with 15mM Sodium Azide | |
KIF5B | |
Stalk domain of human kinesin (amino acids 331-906) expressed in E. coli (FKHC3). | |
0.1 mg | |
Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
3799 | |
Human, Mouse, Pig | |
Purified |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.0 mg/mL | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
This detects total level of endogenous kinesin protein | |
Store at 4C. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction