Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5B Antibody (KN-03)

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500531
Description
Kinesin 5B Monoclonal specifically detects Kinesin 5B in Human, Mouse, Rat, Porcine samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| Kinesin 5B | |
| Monoclonal | |
| 1.0 mg/mL | |
| Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
| KIF5B | |
| Enriched fraction of porcine brain kinesin. | |
| 0.1 mg | |
| Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
| 3799 | |
| Human, Mouse, Rat, Pig | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| KN-03 | |
| Unconjugated | |
| Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| The antibody Kinesin 5B recognizes heavy chain of conventional kinesin associated with vesicles and with lower affinity with denaturated molecule. It stains Western blots of kinesin-enriched preparations. Epitope mapping (by limited proteolysis of partially purified porcine kinesin) followed by immunoblotting has revealed that antibodies KN-01, KN-02 and KN-03 react with different sets of fragments. The antibody KN-03 well recognizes kinesin bound to taxol-stabilized microtubules. | |
| Store at 4C. Do not freeze. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction