Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131022
Description
Kinesin 5B Polyclonal specifically detects Kinesin 5B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Kinesin 5B | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
KIF5B | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Kinesin 5B. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (83%), Rat (83%), and Xenopus tropicalis (83%). Not yet tested in any other species. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.4mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
Rabbit | |
110 kDa | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience | |
3799 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction