Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kinesin 5B Polyclonal Antibody, Biotin
GREENER_CHOICE

Catalog No. PIPA5142007
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142007 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142007 Supplier Invitrogen™ Supplier No. PA5142007
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.

Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin 5B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene KIF5B
Gene Accession No. P33176
Gene Alias AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745)
Gene Symbols KIF5B
Host Species Goat
Immunogen Peptide with sequence CQPVAVRGGGGKQV (aa 951-963).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100855651, 3799
Target Species Human, Canine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.