Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5B Rabbit anti-Human, Mouse, Rat, Clone: S04-6G8, Novus Biologicals™

Rabbit Monoclonal Antibody
$266.02 - $556.36
Specifications
| Antigen | Kinesin 5B |
|---|---|
| Clone | S04-6G8 |
| Dilution | Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:20-1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence 20 ug/ml |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Monoclonal |
Description
Kinesin 5B Monoclonal antibody specifically detects Kinesin 5B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
| Kinesin 5B | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry 1:20-1:100, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence 20 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Conventional kinesin heavy chain, kinesin family member 5B, kinesin-1 heavy chain, KNS1kinesin 1 (110-120kD), KNSKINH, Ubiquitous kinesin heavy chain, UKHCkinesin heavy chain | |
| A synthetic peptide of human Kinesin 5B (Uniprot#: P33176) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S04-6G8 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 3799 | |
| IgG | |
| Affinity purified | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title