Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kinesin 5B Recombinant Rabbit Monoclonal Antibody (JE64-16)
GREENER_CHOICE

Catalog No. PIMA544763
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544763 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544763 Supplier Invitrogen™ Supplier No. MA544763
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Kinesin 5B Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Microtubule-dependent motor required for normal distribution of mitochondria and lysosomes. Can induce formation of neurite-like membrane protrusions in non-neuronal cells in a ZFYVE27-dependent manner. Regulates centrosome and nuclear positioning during mitotic entry. During the G2 phase of the cell cycle in a BICD2-dependent manner, antagonizes dynein function and drives the separation of nuclei and centrosomes. Required for anterograde axonal transportation of MAPK8IP3/JIP3 which is essential for MAPK8IP3/JIP3 function in axon elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin 5B
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE64-16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene KIF5B
Gene Accession No. P33176, Q2PQA9, Q61768
Gene Alias AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745)
Gene Symbols KIF5B
Host Species Rabbit
Immunogen Synthetic peptide within human KIF5B aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117550, 16573, 3799
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.