Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kinesin 5C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5116217
Description
Kinesin 5C Polyclonal Antibody for Western Blot
Kinesins are a superfamily of microtubule-associated motor proteins involved in a variety of cellular processes including membranous organelle transport and cell division. Kinesin has been found in a variety of organisms and cell types and is subject to spatial and temporal regulation. These proteins have a modular structure including a conserved motor domain of approximately 350 amino acids, which is responsible for microtubule binding and ATP hydrolysis. In addition to the motor domain, subfamily members share common domain organization, exhibit sequence similarity, motility properties, and cellular functions outside of the motor domain. There are currently three known Kinesin 5 family members denoted as A, B, and C. Kinesin 5A and kinesin 5C appear to be exclusively neuronal, whereas kinesin 5B appears to be ubiquitous in its expression.
Specifications
Kinesin 5C | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
O60282, P28738 | |
KIF5C | |
KLH conjugated synthetic peptide between 758-786 amino acids from the C-terminal region of human KIF5C. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KIF5C | |
CDCBM2; Khc; KIAA0531; KIF5C; kinesin family member 5C; kinesin heavy chain isoform 5C; kinesin heavy chain member 5C; kinesin heavy chain neuron-specific 2; KINN; NKHC; NKHC2; NKHC-2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
16574, 3800 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction