Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin 5C Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Kinesin 5C |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Kinesin 5C Polyclonal specifically detects Kinesin 5C in Human samples. It is validated for Western Blot.Specifications
Kinesin 5C | |
Polyclonal | |
Purified | |
RUO | |
Human | |
heavy chain, neuron-specific, KIF 5C, kinesin family member 5C, Kinesin heavy chain neuron-specific 2, MGC111478, NKHC, NKHC2NKHC-2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 510-590 of human Kinesin 5C (NP_004513.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
PBS with 50% glycerol, pH7.3. | |
3800 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title