Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin C2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15824720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15824720 20 μL
NBP158247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15824720 Supplier Novus Biologicals Supplier No. NBP15824720UL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

Kinesin C2 Polyclonal specifically detects Kinesin C2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Kinesin C2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q96AC6
Gene Alias kinesin family member C2, kinesin-like protein KIFC2
Gene Symbols KIFC2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to KIFC2 (kinesin family member C2) The peptide sequence was selected from the N terminal of KIFC2. Peptide sequence VRPPSPDGSTSQEESPSHFTAVPGEPLGDETQGQQPLQLEEDQRAWQRLE.
Purification Method Protein A purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cytoskeleton Markers, Vision
Primary or Secondary Primary
Gene ID (Entrez) 90990
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.