Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin Light Chain, Mouse anti-Rat, Mouse, Squid, Clone: 63-90, Millipore Sigma™

Catalog No. MABT148925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT148925 25 μg
MABT1489100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT148925 Supplier MilliporeSigma Supplier No. MABT148925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Kinesin Light Chain, clone 63-90, Cat. No. MABT1489, is a mouse monoclonal antibody that detects Kinesin light chain and has been tested for use in Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.

Kinesin light chain (UniProt: P37285 and B2GV74 for KLC1 and KLC2, respectively) is encoded by the Klc1/Klc2 (also known as Klc, Kns2) gene (Gene ID: 171041/309159) in rat. Kinesin is a microtubule-associated force-producing protein that may play a role in organelle transport. The kinesin light chain (KLC) may function in coupling of cargo to the heavy chain or in the modulation of its ATPase activity. Kinesin light chain 1 is expressed in the brain tissue and exists as an oligomeric complex of two heavy and two light chains. It also contains six tetratricopeptide repeats (TPR). The heavy chains of kinesin have a microtubule-binding ATPase motor domain at their amino terminus, a neck coil, and an extended series of coiled coils, separated by a hinge region, that results in heavy-chain dimerization. Kinesin light chains can associate with the heavy-chain coiled coils at the carboxyl-terminal region. The light chains bind cargoes and regulate its activity. KLCs through their TRP repeat domains can recognize shirt linear peptide motifs found on cargo proteins. In the absence of any cargo binding, kinesin-1 exists in a folded compact state, which prevents unnecessary cycles of ATP hydrolysis. However, in the cargo-bound active state inhibition is relieved and it adopts a more elongated structure that can hydrolyze ATP and translocate along microtubules. Point mutations in Klc gene can affect functional domains in kinesin, which can result in late-onset dying-back neuropathies in sensory or motor neurons. (Ref.: Yip, YY., et al. (2016). Proc. Natl. Acad. Sci. USA 113(9); 2418-2423; Pigino, G., et al. (2009). Proc. Natl. Acad. Sci. USA 106(14); 5907-5912).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin Light Chain
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 63-90
Conjugate Unconjugated
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution from a representative lot detected Kinesin Light Chain in rat cerebral cortex tissue sections.Western Blotting Analysis: 4μg/mL from a representative lot detected Kinesin Light Chain in rat brain tissue lysate.Immunoprecipitation Analysis: A representative lot immunoprecipitated Kinesin Light Chain in Immunoprecipitation applications (Stenoien, D., et. al. (1997). Mol Biol Cell. 8(4):675-89).Immunohistochemistry Analysis: A representative lot detected Kinesin Light Chain in Immunohistochemistry applications (Rahman, A., et. al. (1998). J Biol Chem. 273(25):15395-403).Western Blotting Analysis: A representative lot detected Kinesin Light Chain in Western Blotting applications (Rahman, A., et. al. (1998). J Biol Chem. 273(25):15395-403; Stenoien, D., et. al. (1997). Mol Biol Cell. 8(4):675-89; Pigino, G., et. al. (2009). Proc Natl Acad Sci U S A. 106(14):5907-12; Pigino, G., et. al. (2003). J Neurosci. 23(11):4499-508).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias KLC1;KLC2;KLC
Gene Symbols Klc1;Klc;Kns2
Host Species Mouse
Immunogen Purified rat brain Kinesin light chain 1.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001075443
Test Specificity Clone 63-90 detects both KLC1 and KLC2. It targets an epitope with in the N-terminal region.
Target Species Rat, Mouse, Squid
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.