Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kinesin light chain Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144359
Description
Kinesin light chain Polyclonal Antibody for Western Blot, ELISA
Kinesin is a microtubule-associated force-producing protein that play a role in organelle transport.
Specifications
| Kinesin light chain | |
| Polyclonal | |
| Unconjugated | |
| Uncharacterized protein | |
| Recombinant Human Uncharacterized protein (502-717AA). | |
| 100 μL | |
| Primary | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction