Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Kinesore
GREENER_CHOICESDP

Catalog No. 666450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-645-0 50 mg
66-641-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 66-645-0 Supplier Tocris Bioscience™ Supplier No. 6664/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kinesin-1 modulator; cell permeable

Kinesore is a kinesin-1 modulator. Inhibits SKIP-KLC2 interaction, but promotes kinesin-1 function in controlling the organization of the microtubule network. Induces remodeling of the microtubule network and the formation of extensive microtubule-rich projections. Cell permeable.

Specifications

Chemical Name or Material 3,5-Dibromo-N'-((2,5-dimethyl-1-(3-nitrophenyl)-1H-pyrrol-3-yl)methylene)-4-hydroxybenzohydrazide
CAS 363571-83-9
Quantity 50 mg
Molecular Formula C20H16Br2N4O4
Formula Weight 536.17
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.